Order Entry
Export
ContactUsLinkComponent
Paraffin, liquid, GPR RECTAPUR®
Paraffin, liquid, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Paraffin, liquid, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Polycyclic aromatic hydrocarbons:
    Passes test
  • Readily carbonisable substances:
    Passes test
  • Solid paraffins:
    Passes test
  • Acidity:
    Max. 0.002 meq/g
  • Colouration:
    Max. 20 APHA
  • Density (20/4):
    0.850 - 0.885
  • Viscosity (40°C):
    Min. 34.5 cSt
  • Ignition residue (SO4):
    Max. 0.1 %

Specification Test Results