Order Entry
Export
ContactUsLinkComponent
 
Paraffin, highly liquid, Sigma-Aldrich®
Catalog # SIAL76233-1L
Supplier:  Merck
CAS Number:  
Paraffin, highly liquid, Sigma-Aldrich®
Catalog # SIAL76233-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Boiling Pt:300…450 °C (1013 hPa)
Density:0.86 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Flash Pt:195 °C
EINECS:232-384-2
CAS Number:8012-95-1
 :64742-51-4
MDL Number:MFCD00131611