Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Paraffin
Paraffin, highly liquid
Paraffin, highly liquid
Paraffin, highly liquid
Supplier:
Roth Carl
Certificates
Expand image
Paraffin, highly liquid
Supplier:
Roth Carl
About this item
CAS Number:
8012-95-1
:
64742-51-4
MDL Number:
MFCD00131611
Boiling Pt:
300…450 °C (1013 hPa)
Density:
0.86 g/cm³ (20 °C)
Flash Pt:
195 °C
EINECS:
232-384-2