Order Entry
Export
ContactUsLinkComponent
Paraffin, highly liquid, GPR RECTAPUR®
Paraffin, highly liquid, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Paraffin, highly liquid, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Density (20/20):
    0.818 - 0.875
  • Viscosity (40°C):
    Max. 33.5 cSt

Specification Test Results