Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Molecular sieve 13X (1.0 nm, 10 Å)
Molecular sieve 13X (1.0 nm, 10 Å), powder
Molecular sieve 13X (1.0 nm, 10 Å), powder
Molecular sieve 13X (1.0 nm, 10 Å), powder
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Molecular sieve 13X (1.0 nm, 10 Å), powder
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
63231-69-6
:
1318-02-1
MDL Number:
MFCD00164506
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAA10378
Documentation