Order Entry
Export
Orders LinkContactUsLinkComponent
Soda lime

Soda lime

Corrosive
Supplier:   Roth Carl
Soda lime
Supplier:  Roth Carl
Certificates

2 Options Available Below

Product Details & Documents
  • CAS Number:8006-28-8
  • MDL Number:MFCD00134094
  • Merck Index:13,08640
  • UN:1907
  • ADR:8,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 51,80 €
    51,80 €
    1 * 1 L
       51,80 €

  • 5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 161,00 €
    161,00 €
    1 * 5 L
       161,00 €