Paraffin, Solidification point: 44-46°C, Sigma-Aldrich®
Catalog # SIAL76228-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
EINECS:232-315-6
CAS Number:8002-74-2
:64742-51-4