0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1,4-Cyclohexanediol
1,4-Cyclohexanediol
1,4-Cyclohexanediol
1,4-Cyclohexanediol
Supplier:
CHEMPUR
Certificates
Expand image
1,4-Cyclohexanediol
Supplier:
CHEMPUR
About this item
CAS Number:
556-48-9
:
Formula:
C₆H₁₂O₂
MDL Number:
MFCD00001448
:
116.16 g/mol
Boiling Pt:
252 °C (1013 hPa)
Density:
1.17 g/cm³ (21.85 °C)
EINECS:
209-126-2