Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Cyclohexanediol
1,2-Cyclohexanediol
1,2-Cyclohexanediol
1,2-Cyclohexanediol
Supplier:
CHEMPUR
Certificates
Expand image
1,2-Cyclohexanediol
Supplier:
CHEMPUR
About this item
CAS Number:
931-17-9
:
Formula:
C₆H₁₂O₂
MDL Number:
MFCD00003861
:
116.16 g/mol
Boiling Pt:
237 °C (1013 hPa)
Melting Pt:
73 °C
Density:
0.948 g/cm³ (20 °C)
EINECS:
213-229-8