1,3-Cyclooctadiene, Sigma-Aldrich®
Catalog # SIAL29570-10ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:108.18 g/mol
Boiling Pt:143 °C (1013 hPa)
Melting Pt:–53 °C
Density:0.869 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:216-929-1
UN:2520
ADR:3,III
CAS Number:1700-10-3
Formula:C₈H₁₂
MDL Number:MFCD00001751