tert-Butyl-4-methoxyphenol (mixture of isomers), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25013-16-5
:
Formula:C₁₁H₁₆O₂
MDL Number:MFCD01779059
:180.25 g/mol
Merck Index:12,01582
Boiling Pt:264 °C (1013 hPa)
Melting Pt:48…55 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:156 °C
Storage Temperature:Ambient
EINECS:246-563-8