Ruthenium, powder, Sigma-Aldrich®
SIAL545023-1G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
:
:101.07 g/mol
:12,08452
:3900 °C (1013 hPa)
:2310 °C
:12.45 g/cm³ (20 °C)
:231-127-1
:3089
:4.1,III
:M-Clarity™ quality level = MQ100
:7440-18-8
:Ru
:MFCD00011207