
Ruthenium ≥99.95% (metals basis), powder -200 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-18-8
:
Formula:Ru
MDL Number:MFCD00011207
:101.07 g/mol
Merck Index:12,08452
Boiling Pt:3900 °C (1013 hPa)
Melting Pt:2310 °C
Density:12.45 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-127-1
UN:3089
ADR:4.1,III
:GHS02