Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nickel
Nickel ≥99.5% (metals basis), rods, Ø 6.35 mm (0.25 in)
Nickel ≥99.5% (metals basis), rods, Ø 6.35 mm (0.25 in)
:
Thermo Fisher Scientific
Nickel ≥99.5% (metals basis), rods, Ø 6.35 mm (0.25 in)
:
Thermo Fisher Scientific
:
7440-02-0
:
:
Ni
:
MFCD00011137
:
58.69 g/mol
:
12,06582
:
3007 °C (1013 hPa)
:
1453 °C
:
8.9 g/cm³ (20 °C)
:
Ambient
:
231-111-4
:
3089
:
4.1,III
:
GHS08
:
GHS07
externalurl.pdf
msds_ALFA011454