Iridium, Sigma-Aldrich®
Catalog # SIAL449229-7.6G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:7,6 g
- Seq:3206
Specifications
About this item
:
:192.22 g/mol
Boiling Pt:5300 °C (1013 hPa)
Melting Pt:2450 °C
EINECS:231-095-9
CAS Number:7439-88-5
Formula:Ir
MDL Number:MFCD00011062