

Iron ≥98%, spherical powder 1-3 µm
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-89-6
:
Formula:Fe
MDL Number:MFCD00010999
:55.85 g/mol
Boiling Pt:3000 °C (1013 hPa)
Melting Pt:1536 °C
Density:7.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-096-4
UN:3089
ADR:4.1,II
:GHS02
:GHS07