Iron ≥99.95% (metals basis), rods, Ø 7.62 mm (0.30 in)
Catalog # 44918.DQ
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:190 mm
- Seq:1796
Specifications
About this item
CAS Number:7439-89-6
:
Formula:Fe
MDL Number:MFCD00010999
:55.85 g/mol
Merck Index:13,05109
Boiling Pt:3000 °C (1013 hPa)
Melting Pt:1535 °C
Density:7.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-086-4