Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iron
Iron ≥99.99% (metals basis), foil, Thickness: 1.0 mm (0.04 in)
Iron ≥99.99% (metals basis), foil, Thickness: 1.0 mm (0.04 in)
:
Thermo Fisher Scientific
Iron ≥99.99% (metals basis), foil, Thickness: 1.0 mm (0.04 in)
:
Thermo Fisher Scientific
:
7439-89-6
:
:
Fe
:
MFCD00010999
:
55.85 g/mol
:
13,05109
:
3000 °C (1013 hPa)
:
1535 °C
:
7.86 g/cm³ (20 °C)
:
Ambient
:
231-096-4
msds_ALFA040496