Iron ≥99.99% (metals basis), foil, Thickness: 0.1 mm (0.004 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-89-6
:
Formula:Fe
MDL Number:MFCD00010999
:55.85 g/mol
Merck Index:13,05109
Boiling Pt:3000 °C (1013 hPa)
Melting Pt:1535 °C
Density:7.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-096-4