Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Manganese
Manganese ≥99.9% (metals basis), irregular pieces
Manganese ≥99.9% (metals basis), irregular pieces
Manganese ≥99.9% (metals basis), irregular pieces
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Manganese ≥99.9% (metals basis), irregular pieces
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
7439-96-5
:
Formula:
Mn
MDL Number:
MFCD00011111
:
54.94 g/mol
Storage Temperature:
Ambient
EINECS:
231-105-1
Related Information
Documentation
Certificates
Search by Lot Number
Product specification
ALFA10236.A7_TDS_EN
SDS
msds_ALFA010236
Videos
Thermo Scientific Acroseal 25 Years
Documentation