Order Entry
Export
ContactUsLinkComponent
Tungsten, Sigma-Aldrich®
Tungsten, Sigma-Aldrich®
Catalog # SIAL356972-18.9G
Supplier:  Merck
CAS Number:  
Tungsten, Sigma-Aldrich®
Catalog # SIAL356972-18.9G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    18,9 g
  • Seq:
    3287

Specifications

About this item

 :
 :183.84 g/mol
Merck Index:12,09945
Boiling Pt:5660 °C (1013 hPa)
Melting Pt:3410 °C
Density:19 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:231-143-9
CAS Number:7440-33-7
Formula:W
MDL Number:MFCD00011461