Molybdenum, Sigma-Aldrich®
Catalog # SIAL357200-25.6G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25,6 g
- Seq:3323
Specifications
About this item
:
Merck Index:12,06317
Boiling Pt:3617 °C (1013 hPa)
Melting Pt:2622 °C
Density:10.28 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:95.94 g/mol
Storage Temperature:Ambient
EINECS:231-107-2
CAS Number:7439-98-7
Formula:Mo
MDL Number:MFCD00003465