Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Vanadium
Vanadium ≥99.8% (metals basis), wire, Ø 0.25 mm (0.01 in)
Vanadium ≥99.8% (metals basis), wire, Ø 0.25 mm (0.01 in)
:
Thermo Fisher Scientific
Vanadium ≥99.8% (metals basis), wire, Ø 0.25 mm (0.01 in)
:
Thermo Fisher Scientific
:
7440-62-2
:
:
V
:
MFCD00011453
:
50.94 g/mol
:
12,10053
:
3477 °C (1013 hPa)
:
1896 °C
:
6.015 g/cm³ (20 °C)
:
Ambient
:
231-171-1
msds_ALFA044105