

Tantalum (50 ppm Nb) ≥99.97% (metals basis), powder, Puratronic® -325 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-25-7
:
Formula:Ta
MDL Number:MFCD00011252
:180.95 g/mol
Density:16.6 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-135-5
UN:3089
ADR:4.1,II
:GHS02
:GHS07