Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tantalum
Tantalum ≥99.9% (metals basis), wire, annealed, Ø 0.05 mm (0.002 in)
Tantalum ≥99.9% (metals basis), wire, annealed, Ø 0.05 mm (0.002 in)
:
Thermo Fisher Scientific
Tantalum ≥99.9% (metals basis), wire, annealed, Ø 0.05 mm (0.002 in)
:
Thermo Fisher Scientific
:
7440-25-7
:
:
Ta
:
MFCD00011252
:
180.95 g/mol
:
12,09223
:
5427 °C (1013 hPa)
:
2996 °C
:
16.69 g/cm³ (20 °C)
:
Ambient
:
231-135-5
msds_ALFA040674