0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Niobium
Niobium ≥99.8% (metals basis), foil, Thickness: 2.0 mm (0.08 in)
Niobium ≥99.8% (metals basis), foil, Thickness: 2.0 mm (0.08 in)
:
Thermo Fisher Scientific
Niobium ≥99.8% (metals basis), foil, Thickness: 2.0 mm (0.08 in)
:
Thermo Fisher Scientific
:
7440-03-1
:
:
Nb
:
MFCD00011126
:
92.91 g/mol
:
3700 °C (1013 hPa)
:
2468 °C
:
8.57 g/cm³ (20 °C)
:
Ambient
:
231-113-5
msds_ALFA043231