Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zirconium
Zirconium, Sigma-Aldrich®
Zirconium, Sigma-Aldrich®
Zirconium, Sigma-Aldrich®
SIAL267724-20G
:
Merck
:
Zirconium, Sigma-Aldrich®
SIAL267724-20G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
20 g
Seq:
3290
:
:
M-Clarity™ quality level = MQ100
:
91.22 g/mol
:
231-176-9
:
7440-67-7
:
Zr
:
MFCD00011303