Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zirconium
Zirconium ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)
Zirconium ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)
:
Thermo Fisher Scientific
Zirconium ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)
:
Thermo Fisher Scientific
:
7440-67-7
:
:
Zr
:
MFCD00011303
:
91.22 g/mol
:
Ambient
:
231-176-9
msds_ALFA000416