Titanium, powder, Sigma-Aldrich®
Catalog # SIAL268496-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:47.88 g/mol
Boiling Pt:3277 °C (1013 hPa)
Melting Pt:1677 °C
Density:4.5 g/cm³ (20 °C)
EINECS:231-142-3
UN:2546
ADR:4.2,II
:M-Clarity™ quality level = MQ100
:GHS02
CAS Number:7440-32-6
Formula:Ti
MDL Number:MFCD00011264