Titanium, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-32-6
:
Formula:Ti
MDL Number:MFCD00011264
:47.88 g/mol
Merck Index:12,09610
Boiling Pt:3262 °C (1013 hPa)
Melting Pt:1660 °C
Density:4.51 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:231-142-3
:GHS02