Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Titanium
Titanium, Sigma-Aldrich®
Titanium, Sigma-Aldrich®
:
Merck
Titanium, Sigma-Aldrich®
:
Merck
:
7440-32-6
:
:
Ti
:
MFCD00011264
:
47.88 g/mol
:
12,09610
:
3262 °C (1013 hPa)
:
1660 °C
:
4.51 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ100
:
231-142-3
:
GHS02