
Titanium, Sigma-Aldrich®
Catalog # SIAL267481-700MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:700 mg
- Seq:2795
Specifications
About this item
:
:47.88 g/mol
Merck Index:12,09610
Boiling Pt:3262 °C (1013 hPa)
Melting Pt:1660 °C
Density:4.51 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:231-142-3
:GHS02
CAS Number:7440-32-6
Formula:Ti
MDL Number:MFCD00011264