Hafnium, Sigma-Aldrich®
Catalog # SIAL266809-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:178.49 g/mol
:M-Clarity™ quality level = MQ100
EINECS:231-166-4
CAS Number:7440-58-6
Formula:Hf
MDL Number:MFCD00011032