Order Entry
Export
ContactUsLinkComponent
Yttrium, Sigma-Aldrich®
Yttrium, Sigma-Aldrich®
Catalog # SIAL262994-5G
Supplier:  Merck
CAS Number:  
Yttrium, Sigma-Aldrich®
Catalog # SIAL262994-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :88.91 g/mol
Merck Index:12,10240
Boiling Pt:2927 °C (1013 hPa)
Melting Pt:1525 °C
Density:4.472 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:231-174-8
CAS Number:7440-65-5
Formula:Y
MDL Number:MFCD00011468