Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Yttrium
Yttrium ≥99.9% (REO, rare earth oxide basis), pieces
Yttrium ≥99.9% (REO, rare earth oxide basis), pieces
:
Thermo Fisher Scientific
Yttrium ≥99.9% (REO, rare earth oxide basis), pieces
:
Thermo Fisher Scientific
:
7440-65-5
:
:
Y
:
MFCD00011468
:
88.91 g/mol
:
12,10240
:
2927 °C (1013 hPa)
:
1525 °C
:
4.472 g/cm³ (20 °C)
:
Packaged under argon.
:
Ambient
:
231-174-8
msds_ALFA000615
Thermo Scientific Acroseal 25 Years