


Cadmium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 2.0 mm (0.08 in)
Catalog # 11367.GH
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
- Pk Info:100×100 mm
Specifications
About this item
:
:112.41 g/mol
Merck Index:12,01649
Boiling Pt:765 °C (1013 hPa)
Melting Pt:321 °C
Density:8.642 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-152-8
UN:2570
ADR:6.1,II
:GHS06
:GHS08
:GHS09
CAS Number:7440-43-9
Formula:Cd
MDL Number:MFCD00010914