Silver, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
- CAS Number:7440-22-4
- Formula:Ag
- MDL Number:MFCD00003397
- Molecular Weight:107.87 g/mol
- Merck Index:12,08647
- Boiling Pt:2212 °C (1013 hPa)
- Melting Pt:962 °C
- Density:10.5 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:231-131-3
2 Options Available Below
Product Details & Documents
- CAS Number:7440-22-4
- Formula:Ag
- MDL Number:MFCD00003397
- Molecular Weight:107.87 g/mol
- Merck Index:12,08647
- Boiling Pt:2212 °C (1013 hPa)
- Melting Pt:962 °C
- Density:10.5 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:231-131-3



