Silver
Glass bottle for solids, 10 g
Catalog # ROTH9373.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:7440-22-4
- Formula:Ag
- MDL Number:MFCD00003397
- Molecular Weight:107.87 g/mol
- Merck Index:12,08647
- Boiling Pt:2212 °C (1013 hPa)
- Melting Pt:962 °C
- Density:10.5 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-131-3
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



