0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Silver
Silver ≥99.95% (metals basis), wire, hard, Ø 0.203 mm (0.008 in)
Silver ≥99.95% (metals basis), wire, hard, Ø 0.203 mm (0.008 in)
:
Thermo Fisher Scientific
Silver ≥99.95% (metals basis), wire, hard, Ø 0.203 mm (0.008 in)
:
Thermo Fisher Scientific
:
7440-22-4
:
:
Ag
:
MFCD00003397
:
107.87 g/mol
:
12,08647
:
2212 °C (1013 hPa)
:
962 °C
:
10.5 g/cm³ (20 °C)
:
Ambient
:
231-131-3
msds_ALFA045071