Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Silver
Silver ≥99.9985% (metals basis), foil, Premion®, Thickness: 1.0 mm (0.04 in)
Silver ≥99.9985% (metals basis), foil, Premion®, Thickness: 1.0 mm (0.04 in)
:
Thermo Fisher Scientific
Silver ≥99.9985% (metals basis), foil, Premion®, Thickness: 1.0 mm (0.04 in)
:
Thermo Fisher Scientific
:
7440-22-4
:
:
Ag
:
MFCD00003397
:
107.87 g/mol
:
12,08647
:
2212 °C (1013 hPa)
:
962 °C
:
10.5 g/cm³ (20 °C)
:
Ambient
:
231-131-3
msds_ALFA012127
Thermo Scientific Acroseal 25 Years