
Copper, powder, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-50-8
:
Formula:Cu
MDL Number:MFCD00010965
:63.55 g/mol
Boiling Pt:2567 °C (1013 hPa)
Melting Pt:1083.4 °C
Density:8.94 g/cm³ (20 °C)
EINECS:231-159-6
ADR:4.1,III
:M-Clarity™ quality level = MQ100
UN:3089
:GHS02
Storage Temperature:Ambient