Order Entry
Export
Orders LinkContactUsLinkComponent
Copper, Sigma-Aldrich®

Copper, Sigma-Aldrich®

Glass bottle for solids, 70 g
Catalog # SIAL349216-70G
Supplier:   Merck
Copper, Sigma-Aldrich®
Catalog # SIAL349216-70G
Supplier:  Merck
CAS Number:  7440-50-8
Copper, Sigma-Aldrich®

Glass bottle for solids, 70 g

CAS Number:  7440-50-8
Price
92,90 €
Log in for availability
1 * 70 g
Product Details & Documents
  • CAS Number:7440-50-8
  • Formula:Cu
  • MDL Number:MFCD00010965
  • Molecular Weight:63.55 g/mol
  • Merck Index:13,02545
  • Boiling Pt:2595 °C (1013 hPa)
  • Melting Pt:1083 °C
  • Density:8.94 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:231-159-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    70 g

Specifications