Copper, Sigma-Aldrich®
Glass bottle for solids, 5,8 g
Catalog # SIAL326410-5.8G
Supplier: Merck
Jump to:
Product Details & Documents
- CAS Number:7440-50-8
- Formula:Cu
- MDL Number:MFCD00010965
- Molecular Weight:63.55 g/mol
- Merck Index:13,02545
- Boiling Pt:2595 °C (1013 hPa)
- Melting Pt:1083 °C
- Density:8.94 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Storage Temperature:Ambient
- EINECS:231-159-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5,8 g



