Gold, Sigma-Aldrich®
Catalog # SIAL349283-150MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:150 mg
- Seq:2500
Specifications
About this item
:
:196.97 g/mol
Merck Index:12,04526
Boiling Pt:2800 °C (1013 hPa)
Melting Pt:1063 °C
Density:19.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-165-9
CAS Number:7440-57-5
Formula:Au
MDL Number:MFCD00003436