Order Entry
Export
ContactUsLinkComponent
Gold ≥99.9% (metals basis), ultrathin foil, Thickness: 1.0 µm
Gold ≥99.9% (metals basis), ultrathin foil, Thickness: 1.0 µm
Catalog # 43149.FF
CAS Number:  
Gold ≥99.9% (metals basis), ultrathin foil, Thickness: 1.0 µm
Catalog # 43149.FF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1643
  • Pk Info:
    25×25 mm

Specifications

About this item

 :
 :196.97 g/mol
Merck Index:12,04526
Boiling Pt:2800 °C (1013 hPa)
Melting Pt:1063 °C
Density:19.3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-165-9
CAS Number:7440-57-5
Formula:Au
MDL Number:MFCD00003436