Terbium ≥99.9% (metals basis), rods, Ø 12.7 mm (0.50 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-27-9
:
Formula:Tb
MDL Number:MFCD00011256
:158.93 g/mol
Merck Index:12,09300
Boiling Pt:3041 °C (1013 hPa)
Melting Pt:1356 °C
Density:8.27 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-137-6