Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Neodymium
Neodymium, Sigma-Aldrich®
Neodymium, Sigma-Aldrich®
SIAL263141-25G
:
Merck
:
Neodymium, Sigma-Aldrich®
SIAL263141-25G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
144.24 g/mol
:
M-Clarity™ quality level = MQ100
:
231-109-3
:
7440-00-8
:
Nd
:
MFCD00011130