Neodymium ≥99.5% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-00-8
:
Formula:Nd
MDL Number:MFCD00011130
:144.24 g/mol
Storage Temperature:Ambient
EINECS:231-109-3