Order Entry
Export
Orders LinkContactUsLinkComponent
Neodymium ≥99.1% (metals basis), ingot

Neodymium ≥99.1% (metals basis), ingot

Neodymium ≥99.1% (metals basis), ingot
Certificates
Neodymium ≥99.1% (metals basis), ingot
Neodymium ≥99.1% (metals basis), ingot

2 Options Available Below

Product Details & Documents

Purity: 99.1% CAS No.: 7440-00-8 Physical Form: Packaged under mineral oil Appearance: Grey/silver MDL No.: MFCD00011130

  • CAS Number:7440-00-8
  • Formula:Nd
  • MDL Number:MFCD00011130
  • Molecular Weight:144.24 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-109-3
Product Family Options

Product Information