Order Entry
Export
ContactUsLinkComponent
Sulphur ≥99.5%, AnalaR NORMAPUR® analytical reagent
Irritant
Sulphur ≥99.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 28260.234
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Sulphur ≥99.5%, AnalaR NORMAPUR® analytical reagent
Catalog # 28260.234
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Acidity or alkalinity:
    Max. 0.002 meq/g
  • Ignition residue:
    Max. 0.04 %
  • Cl (Chloride):
    Max. 50 ppm
  • S (Sulphide):
    Max. 2 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • As (Arsenic):
    Max. 5 ppm

Specification Test Results

About this item

 :
 :32.07 g/mol
Merck Index:13,09059
Boiling Pt:445 °C (1013 hPa)
Melting Pt:113…119 °C
Density:2.36 g/cm³ (20 °C)
Flash Pt:168 °C
Storage Temperature:Ambient
EINECS:231-722-6
UN:1350
ADR:4.1,III
 :GHS07
CAS Number:7704-34-9
Formula:S
MDL Number:MFCD00085316